Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,4-triazin-5(2H)-one, 3,4-dihydro-2-β-D-ribofuranosyl-3-thioxo- | 27089-56-1 | MFCD00006473 | 97.0% | 261.26 g/mol | C8H11N3O5S | 50 MG
SDP

Catalog No. 5000299815
Encompass_Preferred

2-Thio-6-azauridine is a biochemical assay reagent used in research applications. It is a thio analog of 6-azauridine (C8H11N3O5S) with a molecular weight of 261.26 g/mol, supplied as a high-purity solid for laboratory assays. Store under recommended conditions and use for research purposes only.

  • High purity (97.0%).
  • Molecular formula C8H11N3O5S; molecular weight 261.26 g/mol.
  • Intended for biochemical assays and research use only.
  • Storage: solid at 4°C; in solvent, store under nitrogen at -80°C (6 months) or -20°C (1 month).
  • Supplied as a solid for convenient handling in laboratory workflows.

Catalog No. 50-002-99815 Supplier Medchemexpress LLC Supplier No. HYW03927250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

2-Thio-6-azauridine is a biochemical assay reagent used in research applications. It is a thio analog of 6-azauridine (C8H11N3O5S) with a molecular weight of 261.26 g/mol, supplied as a high-purity solid for laboratory assays. Store under recommended conditions and use for research purposes only.

  • High purity (97.0%).
  • Molecular formula C8H11N3O5S; molecular weight 261.26 g/mol.
  • Intended for biochemical assays and research use only.
  • Storage: solid at 4°C; in solvent, store under nitrogen at -80°C (6 months) or -20°C (1 month).
  • Supplied as a solid for convenient handling in laboratory workflows.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.