Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Triazole-3-carboxamide 97.0+%, TCI America™
SDP

Catalog No. T24025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T24025G 5 g
T240225G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T24025G Supplier TCI America Supplier No. T24025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 8-5-3641
Molecular Formula C3H4N4O
Molecular Weight (g/mol) 112.09
MDL Number MFCD03990481
InChI Key ZEWJFUNFEABPGL-UHFFFAOYSA-N
PubChem CID 65125
IUPAC Name 1H-1,2,4-triazole-5-carboxamide
SMILES C1=NNC(=N1)C(=O)N

Specifications

Melting Point 315°C
Color White-Yellow
Quantity 5 g
Formula Weight 112.09
Percent Purity ≥97.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,4-Triazole-3-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.