Learn More
Medchemexpress LLC 1,2,4-triazole-3-carboxylic acid | 4928-87-4 | MFCD00078268 | 97.0% | 113.07 g/mol | C3H3N3O2 | 100 G
SDP
Catalog No. 5000240288
Encompass_Preferred
1,2,4-Triazole-3-carboxylic acid (CAS 4928-87-4) is a small heterocyclic carboxylic acid supplied as a solid reagent for research and analytical applications. Typical purity is 97%, the molecular formula is C3H3N3O2, and the molecular weight is 113.07 g/mol. The material is intended for laboratory research use only and has documented storage recommendations for solid and solution forms.
  • 97% purity suitable for analytical and assay use.
  • Small heterocyclic carboxylic acid useful as a synthetic building block.
  • Available in multiple pack sizes from 1 g to 100 g.
  • Stable at room temperature as a solid; in solution, store at -80 °C or -20 °C per guidelines.
  • Molecular weight 113.07 g/mol, molecular formula C3H3N3O2.
Catalog No. 50-002-40288 Supplier Medchemexpress LLC Supplier No. HY107870100G
1,2,4-Triazole-3-carboxylic acid (CAS 4928-87-4) is a small heterocyclic carboxylic acid supplied as a solid reagent for research and analytical applications. Typical purity is 97%, the molecular formula is C3H3N3O2, and the molecular weight is 113.07 g/mol. The material is intended for laboratory research use only and has documented storage recommendations for solid and solution forms.
  • 97% purity suitable for analytical and assay use.
  • Small heterocyclic carboxylic acid useful as a synthetic building block.
  • Available in multiple pack sizes from 1 g to 100 g.
  • Stable at room temperature as a solid; in solution, store at -80 °C or -20 °C per guidelines.
  • Molecular weight 113.07 g/mol, molecular formula C3H3N3O2.