Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2,4-triazole-3-carboxylic acid | 4928-87-4 | MFCD00078268 | 97.0% | 113.07 g/mol | C3H3N3O2 | 25 G
SDP

Catalog No. 5000242434
Encompass_Preferred

1,2,4-Triazole-3-carboxylic acid is a research-grade small-molecule biochemical reagent supplied at 97% purity (CAS 4928-87-4). It serves as a reagent or intermediate in biochemical assays and synthetic chemistry. The compound is provided as a solid and is stable when stored under the recommended conditions, with documented storage stability both at room temperature and when frozen in solution for extended retention.

  • High purity suitable for research use.
  • Molecular formula C3H3N3O2 and molecular weight 113.07 g/mol.
  • Solid form for easy handling and weighing.
  • Recommended storage: room temperature for routine storage; freeze in solvent for extended stability.
  • Intended for biochemical assay and research applications only.

Catalog No. 50-002-42434 Supplier Medchemexpress LLC Supplier No. HY10787025G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1,2,4-Triazole-3-carboxylic acid is a research-grade small-molecule biochemical reagent supplied at 97% purity (CAS 4928-87-4). It serves as a reagent or intermediate in biochemical assays and synthetic chemistry. The compound is provided as a solid and is stable when stored under the recommended conditions, with documented storage stability both at room temperature and when frozen in solution for extended retention.

  • High purity suitable for research use.
  • Molecular formula C3H3N3O2 and molecular weight 113.07 g/mol.
  • Solid form for easy handling and weighing.
  • Recommended storage: room temperature for routine storage; freeze in solvent for extended stability.
  • Intended for biochemical assay and research applications only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.