Learn More
Medchemexpress LLC 1,2,4-triazolo[4,3-b]pyridazine, 3-[(2-methyl-2H-indazol-5-yl)thio]-6-(1-methyl-1H-pyrazol-4-yl)- | 1357072-61-7 | MFCD26960936 | 98.9% | 362.41 g/mol | C17H14N8S | 10 MG
SDP
Catalog No. 5000288411
Encompass_Preferred
c-Met inhibitor 1 is an orally active small-molecule inhibitor of the c-Met (HGFR) receptor tyrosine kinase with reported antitumor activity. It is supplied as a research-grade solid suitable for in vitro and in vivo pharmacology studies.
  • Orally active c-Met kinase inhibitor for preclinical research.
  • Demonstrated antitumor activity in multiple xenograft models.
  • High purity, 98.9% by supplier analysis.
  • Solid powder form suitable for formulation and dosing studies.
  • Identifying data: CAS 1357072-61-7; formula C17H14N8S; MW 362.41 g/mol.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.
Catalog No. 50-002-88411 Supplier Medchemexpress LLC Supplier No. HY1573510MG
c-Met inhibitor 1 is an orally active small-molecule inhibitor of the c-Met (HGFR) receptor tyrosine kinase with reported antitumor activity. It is supplied as a research-grade solid suitable for in vitro and in vivo pharmacology studies.
  • Orally active c-Met kinase inhibitor for preclinical research.
  • Demonstrated antitumor activity in multiple xenograft models.
  • High purity, 98.9% by supplier analysis.
  • Solid powder form suitable for formulation and dosing studies.
  • Identifying data: CAS 1357072-61-7; formula C17H14N8S; MW 362.41 g/mol.
  • Recommended storage: powder at -20°C for long-term, 4°C for short-term.