Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,4-Trivinylcyclohexane (mixture of isomers) 97.0+%, TCI America™
SDP

Catalog No. T0899500ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0899500ML 500 mL
T089925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T0899500ML Supplier TCI America Supplier No. T0899500ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2855-27-8
Molecular Formula C12H18
Molecular Weight (g/mol) 162.28
MDL Number MFCD00001515
InChI Key KTRQRAQRHBLCSQ-UHFFFAOYNA-N
PubChem CID 96529
IUPAC Name 1,2,4-triethenylcyclohexane
SMILES C=CC1CCC(C=C)C(C1)C=C

Specifications

Color Yellow
Boiling Point 88°C
Quantity 500 mL
Formula Weight 162.28
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1,2,4-Trivinylcyclohexane (mixture of isomers)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.