Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5,6,9,10-Hexabromocyclododecane 97.0+%, TCI America™
SDP

Catalog No. H0544500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
500 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H0544500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. H0544500G Supplier TCI America Supplier No. H0544500G

missing translation for 'validMainframeMsg'

Specifications

CAS 3194-55-6
Melting Point 177°C
Molecular Formula C12H18Br6
MDL Number MFCD00003716
Quantity 500 g
Synonym cyclododecane, 1,2,5,6,9,10-hexabromo, hbcd, dsstox_cid_7527, dsstox_rid_78489, dsstox_gsid_27527, saytex hbcd-lm, acmc-1bn89, ksc491m2h, 1,2,5,6,9,10-hexabromocyclododecane, 1,2,5,6,9,10-hexabromocyclododecane 25g
InChI Key DEIGXXQKDWULML-UHFFFAOYSA-N
SMILES C1CC(C(CCC(C(CCC(C1Br)Br)Br)Br)Br)Br
IUPAC Name 1,2,5,6,9,10-hexabromocyclododecane
Molecular Weight (g/mol) 641.7
PubChem CID 18529
Formula Weight 641.70
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,5,6,9,10-Hexabromocyclododecane
Show More Show Less
EINECSNumber (3)-2254
RTECSNumber GU2302500
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.