Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,5-Trimethyl-1H-pyrrole-3-carboxylic acid, Technical Grade, Thermo Scientific™

Catalog No. CC08601DA
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC08601DA 1 g
CC08601CB 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC08601DA Supplier Thermo Scientific Chemicals Supplier No. CC08601DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 175276-50-3
Molecular Formula C8H11NO2
Molecular Weight (g/mol) 153.18
MDL Number MFCD00203866
InChI Key JTEBLTWGSAXWEE-UHFFFAOYSA-N
Synonym 1,2,5-trimethyl-1h-pyrrole-3-carboxylic acid, maybridge1_002499, 3-carboxy-1,2,5-trimethyl-1h-pyrrole, 1,2,5-trimethyl-3-pyrrolecarboxylic acid, 1h-pyrrole-3-carboxylicacid,1,2,5-trimethyl, 1h-pyrrole-3-carboxylicacid, 1,2,5-trimethyl, 1h-pyrrole-3-carboxylic acid, 1,2,5-trimethyl
PubChem CID 736491
IUPAC Name 1,2,5-trimethylpyrrole-3-carboxylic acid
SMILES CN1C(C)=CC(C(O)=O)=C1C

Specifications

Quantity 1 g
Formula Weight 153.18
Percent Purity Tech.
Grade Technical
Chemical Name or Material 1, 2, 5-Trimethyl-1H-pyrrole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.