Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Inline Filter Assembly, 0.5 μm, UPLC, High Pressure

Supplier:  Waters Corp 289002111

Encompass_Preferred

This 0.5 μm inline filter assembly is engineered for high-performance liquid chromatography, specifically designed to integrate with UPLC systems. It effectively filters solvents, ensuring optimal system performance and protection in applications requiring high pressures, including those up to 10,000 psi.

  • Provides 0.5 μm filtration.
  • Designed for UPLC system compatibility.
  • Functions as an inline solvent filter.
  • Supports high-pressure applications up to 10,000 psi.
  • Supplied as one unit per package.
  • Replacement recommended as needed.

Catalog No. 50-791-726


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.