Learn More
Gerstel Gerstel 2.5 mL Headspace Syringe, High-Temperature Plunger, Gas-Tight, Replaceable Needle
SDP
Catalog No. NC1266938
Encompass
The GERSTEL 2.5 mL TriStar HS Syringe is designed for high-performance headspace injection in gas chromatography applications. With features like a high-temperature plunger and replaceable needle, it offers durability and precision for analyzing volatile and semi-volatile compounds. Its gas-tight design ensures reliable sample introduction, and compatibility with GERSTEL 1-2.5 mL Headspace Syringe Holders enhances usability in demanding analytical environments.
Catalog No. NC1266938 Supplier Gerstel Supplier No. 02200132700
The GERSTEL 2.5 mL TriStar HS Syringe is designed for high-performance headspace injection in gas chromatography applications. With features like a high-temperature plunger and replaceable needle, it offers durability and precision for analyzing volatile and semi-volatile compounds. Its gas-tight design ensures reliable sample introduction, and compatibility with GERSTEL 1-2.5 mL Headspace Syringe Holders enhances usability in demanding analytical environments.