Learn More
Strem, An Ascensus Company CAS# 866926-59-2. 500mg. 1-(2,6-Di-i-propylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, min. 97%. MFCD17018754
SDP
Catalog No. 503666
Encompass
CAS# 866926-59-2. 500mg. 1-(2,6-Di-i-propylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, min. 97%. MFCD17018754. Molecular Weight: 384.99. Molecular Formula: C24H33ClN2. Color/form: white to pink solid Strem# 07-4017. http://www.strem.com/catalog/v/07-4017/
Catalog No. 50-366-6 Supplier Strem, An Ascensus Company Supplier No. 074017500MG
CAS# 866926-59-2. 500mg. 1-(2,6-Di-i-propylphenyl)-3-(2,4,6-trimethylphenyl)-4,5-dihydroimidazolium chloride, min. 97%. MFCD17018754. Molecular Weight: 384.99. Molecular Formula: C24H33ClN2. Color/form: white to pink solid Strem# 07-4017. http://www.strem.com/catalog/v/07-4017/