Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2,6-dichlorophenyl)-3-(5-nitrothiophen-3-yl)urea | 672286-03-2 | 99.5% | 332.16 g/mol | C11H7Cl2N3O3S | 250MG
SDP

Catalog No. 5000236094
Encompass_Preferred

DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) for research use. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM, is supplied as a high-purity solid, and is suitable for in vitro and in vivo research where AQP3 modulation is required.

  • Potent and selective AQP3 inhibitor with IC50 ≈ 0.1-0.4 μM.
  • High purity (≈99.5%).
  • Molecular formula C11H7Cl2N3O3S; molecular weight 332.16 g/mol.
  • Soluble in DMSO ≥ 100 mg/mL; clear solution in recommended in vivo formulations at ≥ 2.08 mg/mL.
  • Stable as powder at -20°C for up to 3 years; in solution stable at -80°C for up to 6 months.
  • Intended for research use only in cancer and inflammation/immunology studies.

Catalog No. 50-002-36094 Supplier Medchemexpress LLC Supplier No. HY126073250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DFP00173 is a potent, selective small-molecule inhibitor of aquaporin-3 (AQP3) for research use. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM, is supplied as a high-purity solid, and is suitable for in vitro and in vivo research where AQP3 modulation is required.

  • Potent and selective AQP3 inhibitor with IC50 ≈ 0.1-0.4 μM.
  • High purity (≈99.5%).
  • Molecular formula C11H7Cl2N3O3S; molecular weight 332.16 g/mol.
  • Soluble in DMSO ≥ 100 mg/mL; clear solution in recommended in vivo formulations at ≥ 2.08 mg/mL.
  • Stable as powder at -20°C for up to 3 years; in solution stable at -80°C for up to 6 months.
  • Intended for research use only in cancer and inflammation/immunology studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.