Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-(2,6-dichlorophenyl)-3-(5-nitrothiophen-3-yl)urea | 672286-03-2 | MFCD00121277 | 99.5% | 332.16 | C11H7Cl2N3O3S | 100MG
SDP

Catalog No. 5000236558
Encompass_Preferred

DFP00173 is a potent, selective inhibitor of aquaporin-3 (AQP3) developed for laboratory research. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM and reduces glycerol permeability in human erythrocytes with an IC50 of ~0.2 μM. The compound is supplied as a solid with multiple size options and as a DMSO stock solution for in vitro studies; it is for research use only.

  • potent, selective aquaporin-3 inhibitor.
  • activity: IC50 approximately 0.1-0.4 μM against AQP3.
  • inhibits glycerol permeability in human erythrocytes with IC50 ≈ 0.2 μM.
  • high purity (about 99.5%).
  • molecular weight 332.16 g/mol.
  • available as solid and as 10 mM solution in DMSO.
  • storage: powder at -20 °C long term; stock solutions at -80 °C recommended.
  • intended for laboratory research use only.

Catalog No. 50-002-36558 Supplier Medchemexpress LLC Supplier No. HY126073100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DFP00173 is a potent, selective inhibitor of aquaporin-3 (AQP3) developed for laboratory research. It inhibits mouse and human AQP3 with an IC50 of approximately 0.1-0.4 μM and reduces glycerol permeability in human erythrocytes with an IC50 of ~0.2 μM. The compound is supplied as a solid with multiple size options and as a DMSO stock solution for in vitro studies; it is for research use only.

  • potent, selective aquaporin-3 inhibitor.
  • activity: IC50 approximately 0.1-0.4 μM against AQP3.
  • inhibits glycerol permeability in human erythrocytes with IC50 ≈ 0.2 μM.
  • high purity (about 99.5%).
  • molecular weight 332.16 g/mol.
  • available as solid and as 10 mM solution in DMSO.
  • storage: powder at -20 °C long term; stock solutions at -80 °C recommended.
  • intended for laboratory research use only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.