Learn More
Medchemexpress LLC 1-(2,6-dihydroxyphenyl)ethanone | 699-83-2 | MFCD00002270 | 99.6% | 152.15 g·mol⁻1 | C8H8O3 | 250mg
SDP
Catalog No. 5000269229
Encompass_Preferred
2 6-Dihydroxyacetophenone a polyphenolic derivative of Acetophenone (HY-Y0989) is an orally active mTOR inhibitor 2 6-Dihydroxyacetophenone shows antioxidant activity 2 6-Dihydroxyacetophenone inhibits cell growth and proliferation in CRC cells 2 6-Dihydroxyacetophenone arrests at G0/G1 phase of cell cycle induces apoptosis and suppresses cell migration in CRC cells 2 6-Dihydroxyacetophenone inhibits xanthine oxidase (XOD) with an IC50 of 1 24 mM 2 6-dihydroxyacetophenone improves uric acid metabolism in hyperuricemia mice reduces plasma cholesterol in hypercholesterolemic rats and inhibits lipid accumulation in HFD-induced obese mice 2 6-Dihydroxyacetophenone can be used for the study of colorectal cancer (CRC) hyperuricemia and hypercholesterolemia[1][2][3][4][5][6]
Catalog No. 50-002-69229 Supplier Medchemexpress LLC Supplier No. HYY0106250MG
2 6-Dihydroxyacetophenone a polyphenolic derivative of Acetophenone (HY-Y0989) is an orally active mTOR inhibitor 2 6-Dihydroxyacetophenone shows antioxidant activity 2 6-Dihydroxyacetophenone inhibits cell growth and proliferation in CRC cells 2 6-Dihydroxyacetophenone arrests at G0/G1 phase of cell cycle induces apoptosis and suppresses cell migration in CRC cells 2 6-Dihydroxyacetophenone inhibits xanthine oxidase (XOD) with an IC50 of 1 24 mM 2 6-dihydroxyacetophenone improves uric acid metabolism in hyperuricemia mice reduces plasma cholesterol in hypercholesterolemic rats and inhibits lipid accumulation in HFD-induced obese mice 2 6-Dihydroxyacetophenone can be used for the study of colorectal cancer (CRC) hyperuricemia and hypercholesterolemia[1][2][3][4][5][6]