Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2,6-Hexanetriol

Catalog No. T66206100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T66206100G 100 g
T6620625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T66206100G Supplier Sigma Aldrich Supplier No. T66206100G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1,2,6-hexanetriol has been used as a precursor to synthesize a trivalent linker containing three thiol moieties (triskelion) for the regioselective terminal activation of chitosan.it can be used to prepare:
  • 1,6-hexanediol (key polymer precursor to polyesters) via hydrodeoxygenation process in the presence of rh-reox/sio2 catalyst
  • Tetrahydropyran-2-methanol, a six-membered oxide ring, by reacting with ethylene carbonate

Specifications

CAS 106-69-4
Density 1.109 g/mL (at 25°C (literature))
Boiling Point 178°C (5 mmHg)
Molecular Formula C6H14O3
Linear Formula HO(CH2)4 CH(OH)CH2OH
Refractive Index n20/D 1.476 (literature)
Quantity 100 g
MDL Number MFCD00002976
Synonym 1,2,6-Trihydroxyhexane
Molecular Weight (g/mol) 134.17
Percent Purity 96%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.