Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,6-Hexanetriol, 97+%, extra pure

Catalog No. AC139835000
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle

Catalog No. AC139835000

Supplier: Thermo Scientific Chemicals 139835000

Only null left
Add to Cart

CAS: 106-69-4 | C6H14O3 | 134.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 106-69-4
Molecular Formula C6H14O3
Molecular Weight (g/mol) 134.18
MDL Number MFCD00002976
InChI Key ZWVMLYRJXORSEP-UHFFFAOYNA-N
Synonym 1,2,6-hexanetriol, 1,2,6-trihydroxyhexane, hexanetriol-1,2,6, 1,6-hexanetriol, hexanetriol-1,6, hexane-1,6-triol, 1,6-trihydroxyhexane, acmc-1bsub, dsstox_cid_21224
PubChem CID 7823
IUPAC Name hexane-1,2,6-triol
SMILES OCCCCC(O)CO

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 25.0°C to 32.0°C
Color Yellow
Density 1.1090g/mL
pH 4.0 to 7.0 (30% soln. at 20°C)
Boiling Point 178.0°C (6.7 mmHg)
Flash Point 198°C
Infrared Spectrum Authentic
Assay Percent Range 97% min. (GC)
Packaging Glass bottle
Refractive Index 1.4750 to 1.4770
Linear Formula HO(CH2)4CH(OH)CH2OH
Quantity 500 g
Beilstein 01,IV,2784
Specific Gravity 1.109
Solubility Information Solubility in water: miscible with water. Other solubilities: miscible with acetone and alcohol, immiscible with benzene, toluene, heptane and, diethyl ether, practically insoluble in ether, low solubility in non-polar solvents
Formula Weight 134.18
Percent Purity 97+%
Grade Extra Pure
Physical Form Viscous Liquid
Chemical Name or Material 1, 2, 6-Hexanetriol, 97%
Show More Show Less
Hazard Category HYGROSCOPIC
EINECSNumber 203-424-6
RTECSNumber MO4650000
TSCA TSCA
Recommended Storage the product may solidify at ca. 32°C

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.