Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,6-Trihydroxyhexane, Spectrum™ Chemical
SDP

Catalog No. 18621616 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
25 g
100 g
150 g
500 g
1 kg
20 kg
25 kg
100 kg
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-616 100 kg
18-621-609 25 g
18-621-610 100 g
18-621-611 150 g
18-621-612 500 g
18-621-613 1 kg
18-621-614 20 kg
18-621-615 25 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-621-616 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T2117100KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 106-69-4
Molecular Weight (g/mol) 134.18

Specifications

Packaging Drum
Quantity 100 kg
Percent Purity 96%
Grade Ungraded
Chemical Name or Material 1,2,6-Trihydroxyhexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.