Learn More
Cayman Chemical 1-(2,6-Xylyl)-2-thiourea, Purity: ≥98%, CAS: 6396-76-5, 10 mg
SDP
Catalog No. 502916960
Encompass_Preferred
An analytical reference standard categorized as an anesthetic metabolite; a metabolite of xylazine; has been used as a marker for xylazine doping in horses; intended for research and forensic applications
Catalog No. 50-291-6960 Supplier Cayman Chemical Supplier No. 3937310 MG
An analytical reference standard categorized as an anesthetic metabolite; a metabolite of xylazine; has been used as a marker for xylazine doping in horses; intended for research and forensic applications