Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2,8-Octanetriol 97.0+%, TCI America™
SDP

Catalog No. O02795G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02795G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02795G Supplier TCI America Supplier No. O02795G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 382631-43-8
Molecular Formula C8H18O3
Molecular Weight (g/mol) 162.229
MDL Number MFCD02093484
InChI Key GKCGJDQACNSNBB-UHFFFAOYSA-N
PubChem CID 23431932
IUPAC Name octane-1,2,8-triol
SMILES C(CCCO)CCC(CO)O

Specifications

Melting Point 48°C
Color White
Quantity 5 g
Formula Weight 162.23
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2,8-Octanetriol
EINECSNumber (2)-2391
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.