Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A24955G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24955G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A24955G Supplier TCI America Supplier No. A24955G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 130198-05-9
Molecular Formula C15H24ClNO2
Molecular Weight (g/mol) 285.812
MDL Number MFCD06658143
InChI Key NTKXIDDUCSFBBF-UHFFFAOYSA-N
Synonym 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hydrochloride, 1-2-amino-1-4-methoxyphenyl-ethyl-cyclohexanol hydrochloride, 2-4-methoxyphenyl-2-1-hydroxycyclohexyl ethylamine hydrochloride, 1-4-methoxyphenyl-2-aminoethyl cyclohexanol hydrochloride, n,n-didesmethylvenlafaxine hydrochloride, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexan-1-ol hydrochloride, 1-2-amino-1-4-methyoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol.hcl, 1-2-amino-1-4-methoxyphenyl ethyl cyclohexanol hcl
PubChem CID 9795856
IUPAC Name 1-[2-amino-1-(4-methoxyphenyl)ethyl]cyclohexan-1-ol;hydrochloride
SMILES COC1=CC=C(C=C1)C(CN)C2(CCCCC2)O.Cl

Specifications

Melting Point 176°C
Color White-Yellow
Quantity 5 g
Formula Weight 285.81
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 1-[2-Amino-1-(4-methoxyphenyl)ethyl]cyclohexanol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.