Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-(2-Aminoethyl)piperazine

Catalog No. A55209100G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A55209100G 100 g
A55209500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A55209100G Supplier Sigma Aldrich Supplier No. A55209100G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Utilized in a variety of reactions for studying corrosion inhibition, biological activity, and metal ligand effects on catalysis.

Specifications

CAS 140-31-8
Density 0.985 g/mL (at 25°C (literature))
Boiling Point 218°C to 222°C (lit.)
Molecular Formula C6H15N3
Refractive Index n20/D 1.5 (literature)
Linear Formula C6H15N3
MDL Number MFCD00005971
Quantity 100 g
Synonym 2-Piperazinoethylamine
Molecular Weight (g/mol) 129.2
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.