Alfa Aesar™ 1-(2-Aminoethyl)piperazine, 98%

Manufacturer: Alfa Aesar™ A101540B

Quantity 1000g

 View more versions of this product

Catalog No. AAA101540B

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 140-31-8
    Molecular Formula C6H15N3
    Formula Weight 129.21
    MDL Number MFCD00005971
    UN Number UN2815
    Beilstein 104363
    Synonym 1-Piperazineethylamine; 2-(1-Piperazinyl)ethylamine
    Chemical Name or Material 1- (2-Aminoethyl)piperazine

    Specifications

    Formula Weight 129.21
    Health Hazard 3 GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 1000g
    Flash Point 93°C (199°F)
    CAS 140-31-8
    RTECS Number TK8050000
    TSCA TSCA
    Boiling Point 222°C
    Synonym 1-Piperazineethylamine; 2-(1-Piperazinyl)ethylamine
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: N-AMINOETHYLPIPERAZINE
    Melting Point −19°C
    Molecular Formula C6H15N3
    MDL Number MFCD00005971
    UN Number UN2815
    Beilstein 104363
    Health Hazard 2 GHS H Statement
    H314-H302-H312-H317-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Combustible liquid.
    Chemical Name or Material 1- (2-Aminoethyl)piperazine
    Assay 98%

    Description

    Applications
    1-(2-Aminoethyl)piperazine is utilized in a variety of reactions for studying corrosion inhibition, biological activity and metal ligand effects on catalysis. It is used for epoxy curing, surface activation, and as an asphalt additive. It is used in lube oil and fuel additives, mineral processing aids, polyamide resins, urethane chemicals, wet strength resins.

    Solubility
    Soluble in water.

    Notes
    Air Sensitive. Incompatible with air and oxidizing agents. Store under dry inert gas.

    GHS H Statement
    H314-H302-H312-H317-H227
    Causes severe skin burns and eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Combustible liquid.

    GHS P Statement
    P210-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces.
    - No smoking.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit