Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Azetidinedicarboxylic acid, 1-(1,1-dimethylethyl) ester, (2R) | 228857-58-7 | MFCD01320888 | 99.0% | 201.22 | C9H15NO4 | 500 MG
SDP

Catalog No. 5000300828
Encompass_Preferred

1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected azetidine-derived amino acid used as a synthetic building block and drug intermediate in medicinal chemistry research. Supplied as a white to off-white solid, it is provided with batch analytical data for quality confirmation.

  • CAS registry number: 228857-58-7.
  • Molecular formula: C9H15NO4.
  • Molecular weight: 201.22 g/mol.
  • Purity (HPLC): 98.96%.
  • Appearance: white to off-white solid.
  • Storage and shelf life: store at room temperature, shelf life 3 years; in solvent, -80°C for 2 years or -20°C for 1 year.
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g.
  • Supplied with a certificate of analysis for batch-specific data.

Catalog No. 50-003-00828 Supplier Medchemexpress LLC Supplier No. HYW018751500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Boc-D-azetidine-2-carboxylic acid is a Boc-protected azetidine-derived amino acid used as a synthetic building block and drug intermediate in medicinal chemistry research. Supplied as a white to off-white solid, it is provided with batch analytical data for quality confirmation.

  • CAS registry number: 228857-58-7.
  • Molecular formula: C9H15NO4.
  • Molecular weight: 201.22 g/mol.
  • Purity (HPLC): 98.96%.
  • Appearance: white to off-white solid.
  • Storage and shelf life: store at room temperature, shelf life 3 years; in solvent, -80°C for 2 years or -20°C for 1 year.
  • Available pack sizes: 250 mg, 500 mg, 1 g, 5 g, 10 g, 25 g, 50 g.
  • Supplied with a certificate of analysis for batch-specific data.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.