Learn More
Medchemexpress LLC 1,2-Benzene-3,4,5,6-d4-dicarboxylic acid, mono(phenylmethyl) ester-d4 | 478954-83-5 | 99.35% | 260.28 | 1 MG
SDP
Catalog No. 5000452612
Encompass_Preferred
Monobenzyl phthalate-d4 is the deuterium labeled monobenzyl phthalate, a urinary metabolite found after exposure to phthalates like diethylhexyl phthalate (DEHP). This compound is intended for research use only and can be used as a tracer and an internal standard for quantitative analysis. Deuteration, incorporating stable heavy isotopes like deuterium into drug molecules, is studied for its potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-52612 Supplier Medchemexpress LLC Supplier No. HYW011848S1MG
Monobenzyl phthalate-d4 is the deuterium labeled monobenzyl phthalate, a urinary metabolite found after exposure to phthalates like diethylhexyl phthalate (DEHP). This compound is intended for research use only and can be used as a tracer and an internal standard for quantitative analysis. Deuteration, incorporating stable heavy isotopes like deuterium into drug molecules, is studied for its potential to affect pharmacokinetic and metabolic profiles.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.