Learn More
Medchemexpress LLC 1,2-Benzenediol, 4-methyl- | 452-86-8 | 99.5% | 124.14 | 500 MG
SDP
Catalog No. 5000454868
Encompass_Preferred
4-Methylcatechol is an intermediate in the degradation of certain alkylbenzenes and an orally active suicide inhibitor of catechol 2,3-dioxygenase (C230). It induces apoptosis in melanoma cells through oxidative stress, and some studies suggest it may be carcinogenic. This compound exhibits antiplatelet and blood pressure-lowering activities, and can inhibit protein oxidation in beef.
  • Intermediate in the degradation of certain alkylbenzenes
  • Orally active suicide inhibitor of catechol 2,3-dioxygenase (C23O)
  • Induces apoptosis in melanoma cells via oxidative stress
  • Exhibits antiplatelet activity
  • Exhibits blood pressure-lowering activity
  • Can inhibit protein oxidation in beef
Catalog No. 50-004-54868 Supplier Medchemexpress LLC Supplier No. HYW012814500MG
4-Methylcatechol is an intermediate in the degradation of certain alkylbenzenes and an orally active suicide inhibitor of catechol 2,3-dioxygenase (C230). It induces apoptosis in melanoma cells through oxidative stress, and some studies suggest it may be carcinogenic. This compound exhibits antiplatelet and blood pressure-lowering activities, and can inhibit protein oxidation in beef.
  • Intermediate in the degradation of certain alkylbenzenes
  • Orally active suicide inhibitor of catechol 2,3-dioxygenase (C23O)
  • Induces apoptosis in melanoma cells via oxidative stress
  • Exhibits antiplatelet activity
  • Exhibits blood pressure-lowering activity
  • Can inhibit protein oxidation in beef