Learn More
Medchemexpress LLC 1-[2-(benzhydrylideneamino)oxyethyl]-3,6-dihydro-2H-pyridine-5-carboxylic acid; hydrochloride | 145645-62-1 | MFCD00153853 | 99.8% | 386.87 | C21H23ClN2O3 | 5 MG
SDP
Catalog No. 5000248870
Encompass_Preferred
NNC-711 (NO-711 hydrochloride) is a research-grade GAT-1 (GABA transporter 1) inhibitor used in pharmacology and neuroscience to probe GABA uptake and synaptic inhibition. Supplied as the hydrochloride salt in a solid powder form, it is reported with high purity and defined storage recommendations.
  • Potent, selective GAT-1 inhibitor with low-nanomolar activity for human GAT-1.
  • Hydrochloride salt supplied as a solid powder, suitable for dissolution and formulation.
  • High purity (≈99.8%) for research applications.
  • Defined physical data: molecular weight 386.87, CAS 145645-62-1.
  • Recommended storage: sealed at -20°C; in solvent -80°C up to 6 months.
Catalog No. 50-002-48870 Supplier Medchemexpress LLC Supplier No. HY1035065MG
NNC-711 (NO-711 hydrochloride) is a research-grade GAT-1 (GABA transporter 1) inhibitor used in pharmacology and neuroscience to probe GABA uptake and synaptic inhibition. Supplied as the hydrochloride salt in a solid powder form, it is reported with high purity and defined storage recommendations.
  • Potent, selective GAT-1 inhibitor with low-nanomolar activity for human GAT-1.
  • Hydrochloride salt supplied as a solid powder, suitable for dissolution and formulation.
  • High purity (≈99.8%) for research applications.
  • Defined physical data: molecular weight 386.87, CAS 145645-62-1.
  • Recommended storage: sealed at -20°C; in solvent -80°C up to 6 months.