Learn More
Medchemexpress LLC 1,2-benzisoselenazol-3(2H)-one, 2-[3-(trifluoromethyl)phenyl]- | 2570461-66-2 | 99.4% | 342.17 g·mol⁻¹ | C14H8F3NOSe | 5 MG
SDP
Catalog No. 5000221197
Encompass_Preferred
SARS-CoV-2-IN-7 is a small-molecule inhibitor that reduces SARS-CoV-2 replication in vitro, with a reported IC50 of 844 nM in infected Vero E6 cells. It is supplied for research use and is available as a solid or as DMSO solution in multiple package sizes.
  • Demonstrated antiviral activity in cell culture (IC50 844 nM).
  • High reported chemical purity (99.4%).
  • Available as solid (mg quantities) and 10 mM DMSO solution formats.
  • Molecular weight 342.17 g·mol⁻¹ and formula C14H8F3NOSe.
  • Suitable for in vitro assays and mechanistic studies of viral replication.
Catalog No. 50-002-21197 Supplier Medchemexpress LLC Supplier No. HY1418415MG
SARS-CoV-2-IN-7 is a small-molecule inhibitor that reduces SARS-CoV-2 replication in vitro, with a reported IC50 of 844 nM in infected Vero E6 cells. It is supplied for research use and is available as a solid or as DMSO solution in multiple package sizes.
  • Demonstrated antiviral activity in cell culture (IC50 844 nM).
  • High reported chemical purity (99.4%).
  • Available as solid (mg quantities) and 10 mM DMSO solution formats.
  • Molecular weight 342.17 g·mol⁻¹ and formula C14H8F3NOSe.
  • Suitable for in vitro assays and mechanistic studies of viral replication.