Learn More
Medchemexpress LLC 1,2-benzisothiazol-3(2H)-one, 2-(4-methylphenyl)- | 2514-30-9 | MFCD05856231 | ≥98.0% | 241.31 g/mol | C14H11NOS | 50 MG
SDP
Catalog No. 5000248130
Encompass_Preferred
PBIT (CAS 2514-30-9) is a small-molecule inhibitor of the JARID1/KDM5 family of histone demethylases, reported to selectively inhibit JARID1B (KDM5B). Supplied as a research-grade powder, it is used in epigenetics and chromatin studies to probe lysine demethylase activity. Molecular formula C14H11NOS; molecular weight 241.31 g/mol.
  • Selective inhibitor of JARID1/KDM5 histone demethylases.
  • Suitable for in vitro epigenetics and chromatin regulation assays.
  • Supplied as a powder for research use.
  • Molecular formula C14H11NOS; molecular weight 241.31 g/mol.
  • Store powder at -20°C for long-term stability.
Catalog No. 50-002-48130 Supplier Medchemexpress LLC Supplier No. HY10145150MG
PBIT (CAS 2514-30-9) is a small-molecule inhibitor of the JARID1/KDM5 family of histone demethylases, reported to selectively inhibit JARID1B (KDM5B). Supplied as a research-grade powder, it is used in epigenetics and chromatin studies to probe lysine demethylase activity. Molecular formula C14H11NOS; molecular weight 241.31 g/mol.
  • Selective inhibitor of JARID1/KDM5 histone demethylases.
  • Suitable for in vitro epigenetics and chromatin regulation assays.
  • Supplied as a powder for research use.
  • Molecular formula C14H11NOS; molecular weight 241.31 g/mol.
  • Store powder at -20°C for long-term stability.