Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1,2-Benzisothiazol-3(2H)-one, 2-(4-methylphenyl)- | 2514-30-9 | MFCD05856231 | 99.5% | 241.31 g/mol | C14H11NOS | 100 MG
SDP

Catalog No. 5000245389
Encompass_Preferred

PBIT (2-(4-methylphenyl)-1,2-benzisothiazol-3(2H)-one) is a small-molecule inhibitor of the Jumonji AT-rich interactive domain 1 (JARID1) family of histone demethylases, reported to inhibit JARID1B with an IC50 of about 3 μM. It is provided as a high-purity solid for research applications in epigenetics and enzyme inhibition studies.

  • Selective inhibitor of JARID1 family histone demethylases (JARID1B IC50 ≈ 3 μM)
  • High purity (99.46%)
  • Molecular weight 241.31 g/mol
  • Supplied as a solid with common pack sizes including 100 mg
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Suitable for epigenetics and histone demethylase research

Catalog No. 50-002-45389 Supplier Medchemexpress LLC Supplier No. HY101451100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

PBIT (2-(4-methylphenyl)-1,2-benzisothiazol-3(2H)-one) is a small-molecule inhibitor of the Jumonji AT-rich interactive domain 1 (JARID1) family of histone demethylases, reported to inhibit JARID1B with an IC50 of about 3 μM. It is provided as a high-purity solid for research applications in epigenetics and enzyme inhibition studies.

  • Selective inhibitor of JARID1 family histone demethylases (JARID1B IC50 ≈ 3 μM)
  • High purity (99.46%)
  • Molecular weight 241.31 g/mol
  • Supplied as a solid with common pack sizes including 100 mg
  • Recommended storage: powder -20°C; in solvent -80°C (6 months) or -20°C (1 month)
  • Suitable for epigenetics and histone demethylase research

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.