Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Benzisothiazol-3-one, 97%

Catalog No. AAH6049206
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6049206 5 g
AAH6049214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6049206 Supplier Thermo Scientific Chemicals Supplier No. H6049206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 2634-33-5 | C7H5NOS | 151.183 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is an antimicrobial agent and a pharmaceutical intermediate.

Solubility
Soluble in dichloromethane, dimethyl sulfoxide, methanol.

Notes
Store at room temperature. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2634-33-5
Molecular Formula C7H5NOS
Molecular Weight (g/mol) 151.183
MDL Number MFCD00127753
InChI Key DMSMPAJRVJJAGA-UHFFFAOYSA-N
Synonym 1,2-benzisothiazol-3 2h-one, 1,2-benzisothiazolin-3-one, benzisothiazolone, proxel, benzo d isothiazol-3 2h-one, 1,2-benzisothiazoline-3-one, benzisothiazolinone, proxel pl, benzo d isothiazol-3-one, 2,3-dihydro-1,2-benzothiazol-3-one
PubChem CID 17520
ChEBI CHEBI:167099
IUPAC Name 1,2-benzothiazol-3-one
SMILES C1=CC=C2C(=C1)C(=O)NS2

Specifications

Melting Point 154°C to 158°C
Quantity 5 g
UN Number UN3077
Solubility Information Soluble in dichloromethane,dimethyl sulfoxide,methanol.
Formula Weight 151.19
Percent Purity 97%
Physical Form Crystalline Powder or Powder
Chemical Name or Material 1,2-Benzisothiazol-3-one
Hazard Category H302-H315-H317-H318
Hazard Statement GHS H Statement

H318-H302-H317-H315

Causes serious eye damage.

Harmful if swallowed.

May cause an allergic skin reaction.

Causes skin irritation.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P310-P330-P333+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 220-120-9
RTECSNumber DE4620000
TSCA Yes

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.