Learn More
Medchemexpress LLC 1,2-Benzisothiazole, 3-(1-piperazinyl)- | 87691-87-0 | 99.96% | 219.31 | 100 G
SDP
Catalog No. 5000453100
Encompass_Preferred
Antidepressant agent 10 (Compound 4) is an orally active compound. It has IC50 values of 190 nM for 5-HT1A and 110 nM for 5-HT2. It can be used in the research of mental disorders.
  • Orally active compound.
  • Can be used in the research of mental disorders.
  • Also available as an analytical standard intended for research and analytical applications.
Catalog No. 50-004-53100 Supplier Medchemexpress LLC Supplier No. HYW009754100G
Antidepressant agent 10 (Compound 4) is an orally active compound. It has IC50 values of 190 nM for 5-HT1A and 110 nM for 5-HT2. It can be used in the research of mental disorders.
  • Orally active compound.
  • Can be used in the research of mental disorders.
  • Also available as an analytical standard intended for research and analytical applications.