Learn More
Medchemexpress LLC 1 2-Benzisoxazole 100mg
SDP
Catalog No. 5000205616
Encompass_Preferred
1 2-Benzisoxazole (Indoxazene) is a potential substrate of rabbit liver aldehyde oxidase 1 2-Benzisoxazole was used in the discovery and development of zonisamide (HY-B0124) for the study of epilepsy and Parkinson s disease[1]
Catalog No. 50-002-05616 Supplier Medchemexpress LLC Supplier No. HYW103317100MG
1 2-Benzisoxazole (Indoxazene) is a potential substrate of rabbit liver aldehyde oxidase 1 2-Benzisoxazole was used in the discovery and development of zonisamide (HY-B0124) for the study of epilepsy and Parkinson s disease[1]