Learn More
Medchemexpress LLC 1 2-Benzisoxazole 50mg
SDP
Catalog No. 5000205092
Encompass_Preferred
1 2-Benzisoxazole (Indoxazene) is a potential substrate of rabbit liver aldehyde oxidase 1 2-Benzisoxazole was used in the discovery and development of zonisamide (HY-B0124) for the study of epilepsy and Parkinson s disease[1]
Catalog No. 50-002-05092 Supplier Medchemexpress LLC Supplier No. HYW10331750MG
1 2-Benzisoxazole (Indoxazene) is a potential substrate of rabbit liver aldehyde oxidase 1 2-Benzisoxazole was used in the discovery and development of zonisamide (HY-B0124) for the study of epilepsy and Parkinson s disease[1]