Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Bis(2-Aminophenoxy)ethane-N,N,N',N'-tetraacetic acid

Catalog No. A49265G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A49265G 5 g
A49261G 1 g
A4926250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A49265G Supplier Sigma Aldrich Supplier No. A49265G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

In nature 1,2-Bis(2-Aminophenoxy)ethane-N,N,NG,NG-tetra acetic acid (BAPTA) is a main metabolic product of BAPTA -AM (BAPTA-tetra(actoxymethyl ester), a neuro protective agent in cerebral ischemia, in rats.BAPTA is an intercellular Ca2+ chelator.Used to study morphological and signaling events of Ca2+ deficiency.

Specifications

CAS 85233-19-8
Molecular Formula C22H24N2O10
Linear Formula C22H24N2O10
Quantity 5 g
MDL Number MFCD00036255
Synonym 2,2 -(Ethylenedioxy)dianiline-N,N,N,N-tetraacetic acid; BAPTA; Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid
Molecular Weight (g/mol) 476.43
Percent Purity 98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.