Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1,2-Bis(2-Aminophenoxy)Ethane-N,N,N',N'-Tetraacetic Acid

Catalog No. 1451050GF Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-510-50GF 50 g
14-510-1GF 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14-510-50GF Supplier Sigma Aldrich Supplier No. 1451050GF
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1,2-Bis(2-aminophenoxy)ethane-N,N,N'sa,N'sa-tetraacetic acid is a highly selective calcium chelating reagent. Effect of microinjected calcium-loaded 1,2-bis(2-aminophenoxy)ethane-N,N,N'sa,N'sa-tetraacetic acid (CaBAPTA) on cell-to-cell diffusion of carboxyfluorescein has been studied in staminal hairs of Setcreasea purpurea. It is reported to be a metabolite of 1,2-bis(2-aminophenoxy)ethane-N,N,N'sa,N'sa-tetraacetic acid tetra(acetoxymethyl ester) (BAPTA-AM) in rat urine.Highly selective calcium chelating reagent. Suitable for the spectrophotometric monitoring of Ca2+ levels

Specifications

CAS 85233-19-8
Molecular Formula C22H24N2O10
Linear Formula C22H24N2O10
MDL Number MFCD00036255
Quantity 50 g
Synonym BAPTA; Ethylenedioxybis(o-phenylenenitrilo)tetraacetic acid
Molecular Weight (g/mol) 476.43 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.