Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(2-aminophenoxy)ethane-N,N,N',N'-tetraacetic Acid 90.0+%, TCI America™
SDP

Catalog No. B38951G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B38951G 1 g
B3895100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B38951G Supplier TCI America Supplier No. B38951G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 85233-19-8 | C22H24N2O10 | 476.44 g/mol

Chemical Identifiers

CAS 85233-19-8
Molecular Formula C22H24N2O10
Molecular Weight (g/mol) 476.44
MDL Number MFCD00036255
InChI Key FTEDXVNDVHYDQW-UHFFFAOYSA-N
Synonym bapta, bapeta, 1,2-bis 2-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, ca ii-bapta, 1,2-bis-2-aminophenoxy ethane n,n,n',n'-tetraacetic acid, unii-k22ddw77c0, 1,2-bis o-aminophenoxy ethane-n,n,n',n'-tetraacetic acid, bapta tetrasodium salt hydrate, glycine, n,n'-1,2-ethanediylbis oxy-2,1-phenylene bis n-carboxymethyl, 2,2',2,2'-ethane-1,2-diylbis oxy-2,1-phenylenenitrilo tetraacetic acid
PubChem CID 104751
ChEBI CHEBI:60888
IUPAC Name 2-{[2-(2-{2-[bis(carboxymethyl)amino]phenoxy}ethoxy)phenyl](carboxymethyl)amino}acetic acid
SMILES OC(=O)CN(CC(O)=O)C1=CC=CC=C1OCCOC1=CC=CC=C1N(CC(O)=O)CC(O)=O

Specifications

Melting Point 159°C
Color White
Quantity 1 g
Formula Weight 476.44
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(2-aminophenoxy)ethane-N,N,N′,N′-tetraacetic Acid
RTECSNumber MC0423000
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.