Learn More
Strem, An Ascensus Company CAS# 47253-67-6. 100mg. (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, min.97% (R,R)-Me-DUPHOS. MFCD00142336
SDP
Supplier:  Strem, An Ascensus Company 150096100MG
Encompass
CAS# 47253-67-6. 100mg. (-)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)benzene, min.97% (R,R)-Me-DUPHOS. MFCD00142336. Molecular Weight: 306.37. Molecular Formula: C18H28P2. Color/form: white xtl. Strem# 15-0096. http://www.strem.com/catalog/v/15-0096/
Catalog No. 50-901-09719