Learn More
Strem, An Ascensus Company CAS# 305818-67-1. 500mg. (+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (R,R)-Me-BPE-Rh. MFCD04038741
SDP
Supplier:  Strem, An Ascensus Company 450168500MG
Encompass
CAS# 305818-67-1. 500mg. (+)-1,2-Bis((2R,5R)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (R,R)-Me-BPE-Rh. MFCD04038741. Molecular Weight: 556.21. Molecular Formula: Rh(C8H12)(C14H28P2)+BF4ae. Color/form: red-orange xtl. Strem# 45-0168. http://www.strem.com/catalog/v/45-0168/
Catalog No. 50-901-12085