Learn More
Strem, An Ascensus Company CAS# 142184-30-3. 100mg. (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 98+% (S,S)-Et-DUPHOS-Rh. MFCD00269861
SDP
Supplier:  Strem, An Ascensus Company 450151100MG
Encompass
CAS# 142184-30-3. 100mg. (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) trifluoromethanesulfonate, 98+% (S,S)-Et-DUPHOS-Rh. MFCD00269861. Molecular Weight: 722.63. Molecular Formula: Rh(C8H12)(C22H36P2)+CF3SO3ae. Color/form: orange xtl. Strem# 45-0151. http://www.strem.com/catalog/v/45-0151/
Catalog No. 50-901-12065