Learn More
Strem, An Ascensus Company CAS# 213343-65-8. 100mg. (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (S,S)-Me-BPE-Rh. MFCD04038742
SDP
Catalog No. 5090112087
Encompass
CAS# 213343-65-8. 100mg. (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (S,S)-Me-BPE-Rh. MFCD04038742. Molecular Weight: 556.21. Molecular Formula: Rh(C8H12)(C14H28P2)+BF4ae. Color/form: red-orange xtl. Strem# 45-0169. http://www.strem.com/catalog/v/45-0169/
Catalog No. 50-901-12087 Supplier Strem, An Ascensus Company Supplier No. 450169100MG
CAS# 213343-65-8. 100mg. (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (S,S)-Me-BPE-Rh. MFCD04038742. Molecular Weight: 556.21. Molecular Formula: Rh(C8H12)(C14H28P2)+BF4ae. Color/form: red-orange xtl. Strem# 45-0169. http://www.strem.com/catalog/v/45-0169/