Learn More
Strem, An Ascensus Company CAS# 136779-26-5. 100mg. (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane, 98+% (S,S)-Me-BPE. MFCD01073771
SDP
Supplier:  Strem, An Ascensus Company 150105100MG
Encompass
CAS# 136779-26-5. 100mg. (-)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)ethane, 98+% (S,S)-Me-BPE. MFCD01073771. Molecular Weight: 258.33. Molecular Formula: (C6H12)PCH2CH2P(C6H12). Color/form: colorless to pale-yellow liq. Strem# 15-0105. http://www.strem.com/catalog/v/15-0105/
Catalog No. 50-901-09741