Learn More
Strem, An Ascensus Company CAS# 205064-10-4. 100mg. (+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (S,S)-Me-DUPHOS-Rh. MFCD01862467
SDP
Supplier:  Strem, An Ascensus Company 450159100MG
Encompass
CAS# 205064-10-4. 100mg. (+)-1,2-Bis((2S,5S)-2,5-dimethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 98+% (S,S)-Me-DUPHOS-Rh. MFCD01862467. Molecular Weight: 604.26. Molecular Formula: Rh(C8H12)(C18H28P2)+BF4ae. Color/form: red-orange xtl. Strem# 45-0159. http://www.strem.com/catalog/v/45-0159/
Catalog No. 50-901-12075