Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (+)-1,2-Bis[(2S,5S)-2,5-Diethylphospholano]Benzene

Catalog No. 668486500MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
66-848-6500MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 66-848-6500MG Supplier Sigma Aldrich Supplier No. 668486500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

DuPhos and BPE Ligands: Highly Efficient Privileged LigandsSold in collaboration with Namena Corp. for research purposes only. These compounds were made and sold under license from E.I. du Pont de Nemours and Company, which license does not include the right to use the compounds in producing products for sale in the pharmaceutical field.

Specifications

CAS 948595-09-3
Density 1.294 g/mL at 25°C
Boiling Point 25°C to 32°C
Refractive Index n20/D 1.581
Quantity 100 mg
Synonym (2S,2'S,5S,5'S)-2,2',5,5'-Tetraethyl-1,1'-(o-phenylene)diphospholane; (S,S)-Ethyl-DUPHOS
Molecular Weight (g/mol) 3000 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.