Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol 96.0+%, TCI America™
SDP

Catalog No. B18491G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B18491G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B18491G Supplier TCI America Supplier No. B18491G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 808-12-8
Molecular Formula C28H26O2
Molecular Weight (g/mol) 394.514
MDL Number MFCD00026012
InChI Key YOCSNHRJFCUGEU-UHFFFAOYSA-N
Synonym 1,2-Diphenyl-1,2-di(p-tolyl)-1,2-ethanediol
PubChem CID 4612495
IUPAC Name 1,2-bis(4-methylphenyl)-1,2-diphenylethane-1,2-diol
SMILES CC1=CC=C(C=C1)C(C2=CC=CC=C2)(C(C3=CC=CC=C3)(C4=CC=C(C=C4)C)O)O

Specifications

Melting Point 156°C
Color White
Quantity 1 g
Formula Weight 394.51
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1,2-Bis(4-methylphenyl)-1,2-diphenyl-1,2-ethanediol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.