Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(4-pyridyl)ethane, 97%

Catalog No. p-3756936
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC106470010 1 g Glass bottle
AC106470100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC106470010 Supplier Thermo Scientific Chemicals Supplier No. 106470010
Only null left
Add to Cart

CAS: 4916-57-8 | C12H12N2 | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4916-57-8
Molecular Formula C12H12N2
Molecular Weight (g/mol) 184.24
MDL Number MFCD00006451
InChI Key DQRKTVIJNCVZAX-UHFFFAOYSA-N
Synonym 1,2-bis 4-pyridyl ethane, 1,2-di pyridin-4-yl ethane, 4,4'-ethylenedipyridine, 1,2-di 4-pyridyl ethane, 4,4'-ethane-1,2-diyldipyridine, unii-wo1evc302b, pyridine, 4,4'-1,2-ethanediyl bis, wo1evc302b, chembl70083, 4-2-pyridin-4-ylethyl pyridine
PubChem CID 78630
IUPAC Name 4-(2-pyridin-4-ylethyl)pyridine
SMILES C1=CN=CC=C1CCC2=CC=NC=C2

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 107.0°C to 113.0°C
Color Pink or White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula (C5H4N)CH2CH2(C5H4N)
Quantity 1 g
Formula Weight 184.24
Percent Purity 97%
Physical Form Crystalline Powder or Crystals
Chemical Name or Material 1, 2-Bis(4-pyridyl)ethane
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
EINECSNumber 225-543-2

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.