Learn More
Strem, An Ascensus Company CAS# 28240-69-9. 10g. 1,2-Bis(dichlorophosphino)ethane, min. 97%. MFCD00009609
SDP
Catalog No. 5090109698
Encompass
CAS# 28240-69-9. 10g. 1,2-Bis(dichlorophosphino)ethane, min. 97%. MFCD00009609. Molecular Weight: 231.81. Molecular Formula: Cl2PCH2CH2PCl2. Color/form: colorless to pale yellow liq. Strem# 15-0082. http://www.strem.com/catalog/v/15-0082/
Catalog No. 50-901-09698 Supplier Strem, An Ascensus Company Supplier No. 15008210G
CAS# 28240-69-9. 10g. 1,2-Bis(dichlorophosphino)ethane, min. 97%. MFCD00009609. Molecular Weight: 231.81. Molecular Formula: Cl2PCH2CH2PCl2. Color/form: colorless to pale yellow liq. Strem# 15-0082. http://www.strem.com/catalog/v/15-0082/