Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(dichlorophosphino)ethane, 96%

Catalog No. AAB2109214
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2109214 25 g
AAB2109203 1 g
AAB2109206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2109214 Supplier Thermo Scientific Chemicals Supplier No. B2109214

CAS: 28240-69-9 | C2H4Cl4P2 | 231.802 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28240-69-9
Molecular Formula C2H4Cl4P2
Molecular Weight (g/mol) 231.802
MDL Number MFCD00009609
InChI Key SBWAJHLQMFBNIN-UHFFFAOYSA-N
PubChem CID 119904
IUPAC Name dichloro(2-dichlorophosphanylethyl)phosphane
SMILES C(CP(Cl)Cl)P(Cl)Cl

Specifications

Density 1.54
Boiling Point 70°C (1 mmHg)
Flash Point None
Refractive Index 1.588
Quantity 25 g
UN Number UN3265
Sensitivity Moisture sensitive
Synonym 1,2-bis dichlorophosphino ethane, phosphonous dichloride, 1,2-ethanediylbis, ethylenebis dichlorophosphine, phosphonous dichloride, p,p'-1,2-ethanediylbis, phosphonous dichloride,p,p'-1,2-ethanediylbis, dichloro 2-dichlorophosphanyl ethyl phosphane, ethylenebis phosphonous dichloride, 1,2-bis dichlorophosphine ethane, 1, 2-bis dichlorophosphino ethane, pubchem6472
Formula Weight 231.81
Percent Purity 96%
Chemical Name or Material 1,2-Bis(dichlorophosphino)ethane
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.