Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1,2-Bis(diethylphosphino)ethane, 98%

Catalog No. AA39272MD
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA39272MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA39272MD Supplier Thermo Scientific Chemicals Supplier No. 39272MD

missing translation for 'validMainframeMsg'

CAS: 6411-21-8 | C10H24P2 | 206.25 g/mol

It is used in chemical reaearch, organic Intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6411-21-8
Molecular Formula C10H24P2
Molecular Weight (g/mol) 206.25
MDL Number MFCD00015175
InChI Key MIOCUERTSIJEDP-UHFFFAOYSA-N
Synonym 1,2-bis diethylphosphino ethane, bis 1,2-diethylphosphino ethane, 1,2-bis-diethylphosphino-ethane, acmc-1bbse, ethylenebis diethylphosphine, chembl70363, 1,2 bis diethylphosphino ethane, 1,2-bis-diethylphosphino ethane
PubChem CID 138745
SMILES CCP(CC)CCP(CC)CC

Specifications

Color Colorless
Density 0.884 g/mL
Boiling Point 124°C to 126°C (10 mmHg)
Odor Pungent
Linear Formula (CH3CH2)2PCH2CH2P(CH2CH3)2
Quantity 250 mg
UN Number UN2845
Sensitivity Air Sensitive
Solubility Information It is soluble in organic solvents.
IUPAC Name [2-(diethylphosphanyl)ethyl]diethylphosphane
Formula Weight 206.25
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1,2-Bis(diethylphosphino)ethane
Show More Show Less
Hazard Category H250-H302+H312-H315-H319-H335
Hazard Statement GHS H Statement
H250-H252-H301-H311-H331
Catches fire spontaneously if exposed to air.
Self-heating in large quantities
may catch fire.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P210-P222-P261-P264b-P270-P271-P280-P301+P312-P302+P334-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.2; Packing Group: I; Proper Shipping Name: PYROPHORIC LIQUIDS, ORGANIC, N.O.S.
TSCA No
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.