Learn More
Strem, An Ascensus Company CAS# 23936-60-9. 1g. 1,2-Bis(dimethylphosphino)ethane, 98% DMPE. MFCD00008511
SDP
Catalog No. 5090109710
Encompass
CAS# 23936-60-9. 1g. 1,2-Bis(dimethylphosphino)ethane, 98% DMPE. MFCD00008511. Molecular Weight: 150.14. Molecular Formula: (CH3)2PCH2CH2P(CH3)2. Color/form: colorless liq. Strem# 15-0090. http://www.strem.com/catalog/v/15-0090/
Catalog No. 50-901-09710 Supplier Strem, An Ascensus Company Supplier No. 1500901G
CAS# 23936-60-9. 1g. 1,2-Bis(dimethylphosphino)ethane, 98% DMPE. MFCD00008511. Molecular Weight: 150.14. Molecular Formula: (CH3)2PCH2CH2P(CH3)2. Color/form: colorless liq. Strem# 15-0090. http://www.strem.com/catalog/v/15-0090/