Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich [1,2-Bis(Diphenylphosphino)Ethane]Dichloropalladium(II)

Catalog No. 3452371G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-523-71G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 34-523-71G Supplier Sigma Aldrich Supplier No. 3452371G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

[1,2-Bis(Diphenylphosphino)Ethane]Dichloropalladium(II)

Specifications

CAS 2004-14-0
Molecular Formula C3H9LiOSi
Linear Formula (CH3)3SiOLi
MDL Number MFCD00075322
Quantity 10 g
Synonym PdCl2(dppe)
Molecular Weight (g/mol) 96.13 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.